Assay Details
| Gene Symbol: WDR34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743386.3 | XP_002743432.1 | 2 - 3 | 1024 | 69 | |
| XM_009003574.2 | XP_009001822.1 | 2 - 3 | 1024 | 69 | ||
More Information
Set Membership: |
|

WDR34
Genomic Map