Assay Details
| Gene Symbol: LYRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009283.2 | XP_009007531.1 | 3 - 4 | 431 | 76 | |
| XM_009009284.1 | XP_009007532.1 | 3 - 4 | 425 | 76 | ||
| XM_009009285.1 | XP_009007533.1 | 3 - 4 | 268 | 76 | ||
| XM_017978797.1 | XP_017834286.1 | 3 - 4 | 415 | 76 | ||
More Information
Set Membership: |
|

LYRM1
Genomic Map