Assay Details
| Gene Symbol: RIMS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009000989.2 | XP_008999237.1 | 5 - 6 | 1000 | 61 | |
| XM_017974599.1 | XP_017830088.1 | 5 - 6 | 1000 | 61 | ||
More Information
Set Membership: |
|

RIMS3
Genomic Map