Assay Details
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752826.3 | XP_002752872.1 | 7 - 8 | 3454 | 87 | |
XM_003733817.3 | XP_003733865.1 | 7 - 8 | 3454 | 87 | ||
XM_009004332.2 | XP_009002580.1 | 6 - 7 | 1112 | 87 | ||
XM_009004334.2 | XP_009002582.1 | 6 - 7 | 1112 | 87 | ||
XM_009004335.2 | XP_009002583.1 | 6 - 7 | 1112 | 87 | ||
XM_017976266.1 | XP_017831755.1 | 5 - 6 | 932 | 87 | ||
XM_017976267.1 | XP_017831756.1 | 7 - 8 | 970 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
