Assay Details
| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746080.3 | XP_002746126.1 | 3 - 4 | 3535 | 95 | |
| XM_017970809.1 | XP_017826298.1 | 4 - 5 | 3495 | 95 | ||
More Information
Set Membership: |
|

ZBED9
Genomic Map