Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243140.1 | NP_001230069.1 | 8 - 9 | 667 | 59 | |
| GenBank mRNA | FN433468.1 | - | 8 - 9 | 667 | 59 | |
| KJ628096.1 | - | 8 - 9 | 667 | 59 | ||
More Information
Set Membership: |
|

FSHR
Genomic Map