Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753330.3 | XP_002753376.1 | 6 - 7 | 2126 | 68 | |
| XM_009005246.2 | XP_009003494.1 | 6 - 7 | 2129 | 68 | ||
| GenBank mRNA | KJ922553.1 | - | 5 - 6 | 1269 | 68 | |
More Information
Set Membership: |
|

CYP1A1
Genomic Map