Assay Details
| Gene Symbol: GCDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987376.2 | XP_008985624.1 | 11 - 12 | 1451 | 72 | |
| XM_008987378.1 | XP_008985626.1 | 10 - 11 | 1303 | 72 | ||
| XM_008987379.1 | XP_008985627.1 | 9 - 10 | 1252 | 72 | ||
| XM_017968054.1 | XP_017823543.1 | 9 - 10 | 1340 | 72 | ||
More Information
Set Membership: |
|

GCDH
Genomic Map