Assay Details
| Gene Symbol: CD300A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748703.3 | XP_002748749.1 | 6 - 7 | 1082 | 75 | |
| XM_008997704.1 | XP_008995952.1 | 7 - 8 | 964 | 75 | ||
More Information
Set Membership: |
|

CD300A
Genomic Map