Assay Details
| Gene Symbol: PDZRN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752366.3 | XP_002752412.3 | 5 - 6 | 1280 | 66 | |
| XM_017975991.1 | XP_017831480.1 | 3 - 4 | 689 | 66 | ||
More Information
Set Membership: |
|

PDZRN4
Genomic Map