Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004216.2 | XP_009002464.1 | 4 - 5 | 1189 | 68 | |
| XM_017976208.1 | XP_017831697.1 | 4 - 5 | 1189 | 68 | ||
More Information
Set Membership: |
|

TMEM19
Genomic Map