Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733701.3 | XP_003733749.2 | 2 - 3 | 672 | 68 | |
| XM_017975850.1 | XP_017831339.1 | 2 - 3 | 614 | 68 | ||
| XM_017975851.1 | XP_017831340.1 | 2 - 3 | 511 | 68 | ||
| XM_017975852.1 | XP_017831341.1 | 2 - 3 | 293 | 68 | ||
More Information
Set Membership: |
|

NINJ2
Genomic Map