Assay Details
| Gene Symbol: NXT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002763156.4 | XP_002763202.1 | 3 - 4 | 385 | 90 | |
| XM_008989751.2 | XP_008987999.2 | 3 - 4 | 191 | 90 | ||
| XM_008989752.2 | XP_008988000.1 | 2 - 3 | 165 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008989752.2 transcripts.
Set Membership: |
|

NXT2
Genomic Map