Assay Details
| Gene Symbol: SH3BP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749914.3 | XP_002749960.1 | 4 - 5 | 2729 | 70 | |
| XM_017973811.1 | XP_017829300.1 | 3 - 4 | 2813 | 70 | ||
| XM_017973812.1 | XP_017829301.1 | 3 - 4 | 2855 | 70 | ||
More Information
Set Membership: |
|

SH3BP4
Genomic Map