Assay Details
| Gene Symbol: ARRDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743507.2 | XP_002743553.1 | 1 - 2 | 191 | 100 | |
| XM_009006232.2 | XP_009004480.1 | 1 - 2 | 190 | 100 | ||
More Information
Set Membership: |
|

ARRDC1
Genomic Map