Assay Details
| Gene Symbol: BTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745724.4 | XP_002745770.1 | 2 - 3 | 527 | 59 | |
| XM_008993177.2 | XP_008991425.1 | 2 - 3 | 513 | 59 | ||
| XM_008993178.2 | XP_008991426.1 | 2 - 3 | 687 | 59 | ||
| XM_017970491.1 | XP_017825980.1 | 2 - 3 | 683 | 59 | ||
| XM_017970492.1 | XP_017825981.1 | 2 - 3 | 529 | 59 | ||
More Information
Set Membership: |
|

BTC
Genomic Map