Assay Details
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745439.3 | XP_002745485.1 | 2 - 3 | 753 | 107 | |
XM_008992762.2 | XP_008991010.1 | 2 - 3 | 285 | 107 |

More Information
Set Membership: |
![]() ![]() |
