Assay Details
Gene Symbol: NDNF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745439.3 | XP_002745485.1 | 3 - 4 | 875 | 97 | |
XM_008992762.2 | XP_008991010.1 | 3 - 4 | 407 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
