Assay Details
| Gene Symbol: OSGIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008982867.2 | XP_008981115.1 | 4 - 5 | 777 | 88 | |
| XM_008982868.2 | XP_008981116.1 | 4 - 5 | 777 | 88 | ||
| XM_008982869.2 | XP_008981117.1 | 4 - 5 | 565 | 88 | ||
More Information
Set Membership: |
|

OSGIN2
Genomic Map