Assay Details
| Gene Symbol: FAM192A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966820.1 | XP_017822309.1 | 5 - 6 | 517 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM192A| Gene Symbol: FAM192A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966820.1 | XP_017822309.1 | 5 - 6 | 517 | 65 | |
Set Membership: |
|