Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761678.3 | XP_002761724.1 | 5 | 1449 | 51 | |
| XM_017967636.1 | XP_017823125.1 | 4 | 1383 | 51 | ||
| GenBank mRNA | EF215289.1 | - | 1 | 219 | 51 | |
More Information
Set Membership: |
|

SNAPC2
Genomic Map