Assay Details
Gene Symbol: PTS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754416.4 | XP_002754462.1 | 2 - 3 | 340 | 64 | |
XM_009006811.2 | XP_009005059.1 | 2 - 3 | 363 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
