Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760940.3 | XP_002760986.1 | 3 - 4 | 1814 | 62 | |
| XM_008985851.1 | XP_008984099.1 | 3 - 4 | 466 | 62 | ||
More Information
Set Membership: |
|

SNX20
Genomic Map