Assay Details
| Gene Symbol: NDUFV3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761454.4 | XP_002761500.1 | 1 - 2 | 120 | 85 | |
| XM_008986750.2 | XP_008984998.1 | 1 - 2 | 142 | 85 | ||
More Information
Set Membership: |
|

NDUFV3
Genomic Map