Assay Details
| Gene Symbol: HPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759505.4 | XP_002759551.1 | 13 - 14 | 2610 | 85 | |
| XM_008983678.2 | XP_008981926.1 | 14 - 15 | 2634 | 85 | ||
| XM_008983679.2 | XP_008981927.1 | 14 - 15 | 2634 | 85 | ||
| XM_008983680.2 | XP_008981928.1 | 12 - 13 | 2370 | 85 | ||
| XM_008983681.2 | XP_008981929.1 | 11 - 12 | 2328 | 85 | ||
| XM_017965760.1 | XP_017821249.1 | 14 - 15 | 2391 | 85 | ||
More Information
Set Membership: |
|

HPS3
Genomic Map