Assay Details
| Gene Symbol: CDC42EP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762478.3 | XP_002762524.1 | 3 | 1114 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC42EP5| Gene Symbol: CDC42EP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762478.3 | XP_002762524.1 | 3 | 1114 | 74 | |
Set Membership: |
|