Assay Details
| Gene Symbol: USP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976301.1 | XP_017831790.1 | 3 - 4 | 3031 | 143 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

USP44| Gene Symbol: USP44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976301.1 | XP_017831790.1 | 3 - 4 | 3031 | 143 | |
Set Membership: |
|