Assay Details
| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743815.3 | XP_002743861.1 | 6 - 7 | 1305 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RRP7A| Gene Symbol: RRP7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743815.3 | XP_002743861.1 | 6 - 7 | 1305 | 63 | |
Set Membership: |
|