Assay Details
| Gene Symbol: AGMO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751549.4 | XP_002751595.1 | 3 - 4 | 586 | 62 | |
| XM_009002324.2 | XP_009000572.1 | 3 - 4 | 586 | 62 | ||
| XM_009002328.2 | XP_009000576.1 | 3 - 4 | 586 | 62 | ||
More Information
Set Membership: |
|

AGMO
Genomic Map