Assay Details
Gene Symbol: LY6G5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002806688.3 | XP_002806734.2 | 2 - 3 | 833 | 80 | |
XM_017970969.1 | XP_017826458.1 | 1 - 2 | 983 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
