Assay Details
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008984319.2 | XP_008982567.1 | 3 - 4 | 1054 | 72 | |
XM_008984320.2 | XP_008982568.1 | 3 - 4 | 1054 | 72 | ||
XM_017966255.1 | XP_017821744.1 | 3 - 4 | 1054 | 72 | ||
XM_017966256.1 | XP_017821745.1 | 2 - 3 | 946 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
