Assay Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745331.4 | XP_002745377.1 | 9 - 10 | 925 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF330| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745331.4 | XP_002745377.1 | 9 - 10 | 925 | 64 | |
Set Membership: |
|