Assay Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761891.3 | XP_002761937.1 | 4 - 5 | 1296 | 66 | |
| XM_017968066.1 | XP_017823555.1 | 4 - 5 | 1307 | 66 | ||
More Information
Set Membership: |
|

MVB12A
Genomic Map