Assay Details
| Gene Symbol: PNPLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743834.3 | XP_002743880.1 | 8 - 9 | 1362 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PNPLA3| Gene Symbol: PNPLA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743834.3 | XP_002743880.1 | 8 - 9 | 1362 | 59 | |
Set Membership: |
|