Assay Details
| Gene Symbol: INSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007722.2 | XP_009005970.1 | 8 - 9 | 1121 | 84 | |
| XM_017977928.1 | XP_017833417.1 | 6 - 7 | 2983 | 84 | ||
More Information
Set Membership: |
|

INSC
Genomic Map