Assay Details
| Gene Symbol: DCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744706.4 | XP_002744752.1 | 7 - 8 | 1036 | 60 | |
| XM_003732310.3 | XP_003732358.1 | 7 - 8 | 1036 | 60 | ||
| XM_008991674.2 | XP_008989922.1 | 7 - 8 | 1036 | 60 | ||
| XM_008991675.1 | XP_008989923.1 | 5 - 6 | 745 | 60 | ||
| XM_017969740.1 | XP_017825229.1 | 7 - 8 | 1036 | 60 | ||
More Information
Set Membership: |
|

DCP2
Genomic Map