Assay Details
| Gene Symbol: CCDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762259.3 | XP_002762305.1 | 1 | 2749 | 79 | |
| XM_008988268.2 | XP_008986516.1 | 2 | 2701 | 79 | ||
More Information
Set Membership: |
|

CCDC8
Genomic Map