Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757209.3 | XP_002757255.1 | 5 - 6 | 1079 | 63 | |
| XM_017963452.1 | XP_017818941.1 | 4 - 5 | 2501 | 63 | ||
| XM_017963453.1 | XP_017818942.1 | 4 - 5 | 2507 | 63 | ||
More Information
Set Membership: |
|

ST8SIA5
Genomic Map