Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751483.3 | XP_002751529.1 | 3 - 4 | 531 | 65 | |
| XM_009002278.1 | XP_009000526.1 | 3 - 4 | 531 | 65 | ||
More Information
Set Membership: |
|

FAM221A
Genomic Map