Assay Details
Gene Symbol: FAM107B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008999963.2 | XP_008998211.1 | 2 - 3 | 364 | 86 | |
XM_008999965.2 | XP_008998213.1 | 2 - 3 | 704 | 86 | ||
XM_017974069.1 | XP_017829558.1 | 2 - 3 | 709 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
