Assay Details
| Gene Symbol: SOCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752855.4 | XP_002752901.1 | 2 - 3 | 976 | 93 | |
| XM_009004369.2 | XP_009002617.1 | 2 - 3 | 732 | 93 | ||
| XM_017976279.1 | XP_017831768.1 | 2 - 3 | 615 | 93 | ||
More Information
Set Membership: |
|

SOCS2
Genomic Map