Assay Details
| Gene Symbol: JPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748723.3 | XP_002748769.1 | 1 - 2 | 374 | 95 | |
| XM_017972828.1 | XP_017828317.1 | 1 - 2 | 345 | 95 | ||
More Information
Set Membership: |
|

JPT1
Genomic Map