Assay Details
Gene Symbol: JPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002748723.3 | XP_002748769.1 | 2 - 3 | 514 | 106 | |
XM_017972828.1 | XP_017828317.1 | 2 - 3 | 485 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
