Assay Details
| Gene Symbol: EXOSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002807592.4 | XP_002807638.2 | 1 - 2 | 108 | 63 | |
| XM_017964852.1 | XP_017820341.1 | 1 - 2 | 95 | 63 | ||
More Information
Set Membership: |
|

EXOSC7
Genomic Map