Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994132.2 | XP_008992380.2 | 2 - 3 | 1005 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCF19| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008994132.2 | XP_008992380.2 | 2 - 3 | 1005 | 66 | |
Set Membership: |
|