Assay Details
| Gene Symbol: ZNF543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270907.1 | NP_001257836.1 | 1 - 2 | 366 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270907.1 transcripts.
Set Membership: |
|

ZNF543
Genomic Map