Assay Details
| Gene Symbol: HSD17B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761202.3 | XP_002761248.1 | 2 - 3 | 703 | 97 | |
| GenBank mRNA | AF272010.1 | - | 1 - 2 | 229 | 97 | |
More Information
Set Membership: |
|

HSD17B2
Genomic Map