Assay Details
| Gene Symbol: TMEM219 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009407.2 | XP_009007655.1 | 4 - 5 | 979 | 65 | |
| XM_009009408.2 | XP_009007656.1 | 4 - 5 | 724 | 65 | ||
| XM_009009409.2 | XP_009007657.1 | 4 - 5 | 816 | 65 | ||
| XM_009009410.1 | XP_009007658.1 | 4 - 5 | 1082 | 65 | ||
| XM_009009411.2 | XP_009007659.1 | 4 - 5 | 712 | 65 | ||
More Information
Set Membership: |
|

TMEM219
Genomic Map