Assay Details
| Gene Symbol: RNF126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986860.2 | XP_008985108.1 | 2 - 3 | 280 | 52 | |
| XM_017967673.1 | XP_017823162.1 | 2 - 3 | 280 | 52 | ||
| XM_017967674.1 | XP_017823163.1 | 2 - 3 | 280 | 52 | ||
| XM_017967675.1 | XP_017823164.1 | 2 - 3 | 280 | 52 | ||
| XM_017967676.1 | XP_017823165.1 | 2 - 3 | 280 | 52 | ||
More Information
Set Membership: |
|

RNF126
Genomic Map