Assay Details
| Gene Symbol: LMCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758593.3 | XP_002758639.1 | 4 - 5 | 950 | 59 | |
| XM_008982075.2 | XP_008980323.1 | 4 - 5 | 928 | 59 | ||
| XM_017965044.1 | XP_017820533.1 | 3 - 4 | 760 | 59 | ||
More Information
Set Membership: |
|

LMCD1
Genomic Map